Product Details
- SNP ID
-
rs150531804
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:39399742 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCAAGCACTTTCAGATCAGAGCCC[C/T]GAAGACGAGCAATGGCAATGAGGTT
- Phenotype
-
MIM: 609103
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FBXO33
PubMed Links
Gene Details
- Gene
- FBXO33
- Gene Name
- F-box protein 33
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_203301.3 |
1780 |
Missense Mutation |
CAG,CGG |
Q481R |
NP_976046.1 |
View Full Product Details