Product Details

SNP ID
rs111308983
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:77046842 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGCTTGAGTTGTCAACTATGCA[G/T]AGGTTCCGCCAGTGATGAGGAGCTC
Phenotype
MIM: 606347 MIM: 613134
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PSTPIP1 PubMed Links

Gene Details

Gene
PSTPIP1
Gene Name
proline-serine-threonine phosphatase interacting protein 1
There are no transcripts associated with this gene.

Gene
TSPAN3
Gene Name
tetraspanin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168412.1 1029 Missense Mutation TAT,TCT Y188S NP_001161884.1
NM_005724.5 1029 Missense Mutation TAT,TCT Y252S NP_005715.1
NM_198902.2 1029 Missense Mutation TAT,TCT Y227S NP_944492.1
XM_017021857.1 1029 Missense Mutation TAT,TCT Y213S XP_016877346.1

View Full Product Details