Product Details

SNP ID
rs112194177
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:90903692 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTTCTTTATTAGAAAGAAACTC[A/G]TTCTGTGTTTCTGAGAAAGGATCGT
Phenotype
MIM: 190030 MIM: 600988
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FES PubMed Links

Gene Details

Gene
FES
Gene Name
FES proto-oncogene, tyrosine kinase
There are no transcripts associated with this gene.

Gene
MAN2A2
Gene Name
mannosidase alpha class 2A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320977.1 11 Intron NP_001307906.1
NM_006122.3 11 Intron NP_006113.2
XM_005254910.3 11 Intron XP_005254967.1
XM_011521565.2 11 Intron XP_011519867.1
XM_011521566.2 11 Intron XP_011519868.1
XM_017022184.1 11 UTR 5 XP_016877673.1
XM_017022185.1 11 Intron XP_016877674.1

View Full Product Details