Product Details

SNP ID
rs112240620
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:67826468 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGTGCATCAAGTGCGGCTACTGC[A/G]GCATGTACTTCTCGCCCAACAAGTT
Phenotype
MIM: 611273
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101929076 PubMed Links

Gene Details

Gene
LOC101929076
Gene Name
uncharacterized LOC101929076
There are no transcripts associated with this gene.

Gene
SKOR1
Gene Name
SKI family transcriptional corepressor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258024.1 762 Missense Mutation AGC,GGC S400G NP_001244953.1
XM_017022173.1 762 Missense Mutation AGC,GGC S214G XP_016877662.1

View Full Product Details