Product Details

SNP ID
rs112642431
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40281977 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGGGCTGGGTCGTCGATGCTCGG[C/G]GCTGGCCGCGGGGGCCGGGCGGCCA
Phenotype
MIM: 608110 MIM: 604114
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
ANKRD63 PubMed Links
Additional Information
For this assay, SNP(s) [rs529183681] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ANKRD63
Gene Name
ankyrin repeat domain 63
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190479.2 610 Missense Mutation CCC,GCC P204A NP_001177408.1
Gene
BUB1B-PAK6
Gene Name
BUB1B-PAK6 readthrough
There are no transcripts associated with this gene.

Gene
PAK6
Gene Name
p21 (RAC1) activated kinase 6
There are no transcripts associated with this gene.

Gene
PLCB2
Gene Name
phospholipase C beta 2
There are no transcripts associated with this gene.

View Full Product Details