Product Details

SNP ID
rs113655030
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43774669 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTTGCTCCCAATCTTCTCCCTC[C/T]TGTAAATCCTGATTGGTCAGGCCTT
Phenotype
MIM: 609885 MIM: 602046 MIM: 605054
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ELL3 PubMed Links

Gene Details

Gene
ELL3
Gene Name
elongation factor for RNA polymerase II 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025165.2 1153 Silent Mutation CAA,CAG Q250Q NP_079441.1
Gene
PDIA3
Gene Name
protein disulfide isomerase family A member 3
There are no transcripts associated with this gene.

Gene
SERF2
Gene Name
small EDRK-rich factor 2
There are no transcripts associated with this gene.

View Full Product Details