Product Details

SNP ID
rs113699778
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:64152318 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCTACAGCGAGTTCCACGCGCTG[C/T]ACAAGCGGGTGAGGCGGCGCCGACC
Phenotype
MIM: 123841 MIM: 601272
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPIB PubMed Links

Gene Details

Gene
PPIB
Gene Name
peptidylprolyl isomerase B
There are no transcripts associated with this gene.

Gene
SNX1
Gene Name
sorting nexin 1
There are no transcripts associated with this gene.

Gene
SNX22
Gene Name
sorting nexin 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024798.2 210 Missense Mutation CAC,TAC H51Y NP_079074.2
XM_005254677.3 210 Intron XP_005254734.1
XM_017022581.1 210 Intron XP_016878070.1

View Full Product Details