Product Details
- SNP ID
-
rs113994185
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:72686244 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAGCTAAAATGGCTGAGGAGAGAG[C/T]CGCGACGGTGAGCGCCGAGATTCTC
- Phenotype
-
MIM: 600374
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BBS4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs56368716] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BBS4
- Gene Name
- Bardet-Biedl syndrome 4
- Gene
- HIGD2B
- Gene Name
- HIG1 hypoxia inducible domain family member 2B
There are no transcripts associated with this gene.
View Full Product Details