Product Details

SNP ID
rs114071059
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:77044322 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTGCTCTCTGAAGAACAAGCACT[A/G]GGTAATGCCAACACCTAGCCTCAGC
Phenotype
MIM: 606347 MIM: 613134
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PSTPIP1 PubMed Links

Gene Details

Gene
PSTPIP1
Gene Name
proline-serine-threonine phosphatase interacting protein 1
There are no transcripts associated with this gene.

Gene
TSPAN3
Gene Name
tetraspanin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168412.1 3549 UTR 3 NP_001161884.1
NM_005724.5 3549 UTR 3 NP_005715.1
NM_198902.2 3549 UTR 3 NP_944492.1
XM_017021857.1 3549 Intron XP_016877346.1

View Full Product Details