Product Details

SNP ID
rs114829905
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:64168760 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTAAGGAACTGTAGCAGTAGGGT[A/T]GAGGAGGGAGCCCTGATTCGCTGAT
Phenotype
MIM: 606274 MIM: 123841
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CSNK1G1 PubMed Links

Gene Details

Gene
CSNK1G1
Gene Name
casein kinase 1 gamma 1
There are no transcripts associated with this gene.

Gene
PPIB
Gene Name
peptidylprolyl isomerase B
There are no transcripts associated with this gene.

View Full Product Details