Product Details

SNP ID
rs114833125
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:68207747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCAGGTGACACACCAGGTCCCCT[C/T]CTGGCCTCTGCCCCACCCAGCCCTC
Phenotype
MIM: 606725
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CALML4 PubMed Links

Gene Details

Gene
CALML4
Gene Name
calmodulin like 4
There are no transcripts associated with this gene.

Gene
CLN6
Gene Name
ceroid-lipofuscinosis, neuronal 6, late infantile, variant
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017882.2 1487 UTR 3 NP_060352.1

View Full Product Details