Product Details

SNP ID
rs115097992
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:65384087 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGGGGGATTTGAGCTGGCAGCT[A/G]TCTCCAGGGGTCTCCTCTAGCACCT
Phenotype
MIM: 604184 MIM: 616810
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IGDCC3 PubMed Links

Gene Details

Gene
IGDCC3
Gene Name
immunoglobulin superfamily DCC subclass member 3
There are no transcripts associated with this gene.

Gene
IGDCC4
Gene Name
immunoglobulin superfamily DCC subclass member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020962.2 3692 Silent Mutation GAC,GAT D1225D NP_066013.1
XM_011521845.2 3692 Silent Mutation GAC,GAT D1238D XP_011520147.1
XM_011521846.2 3692 Silent Mutation GAC,GAT D1226D XP_011520148.1
XM_011521847.1 3692 Silent Mutation GAC,GAT D964D XP_011520149.1
XM_017022448.1 3692 Silent Mutation GAC,GAT D1239D XP_016877937.1
XM_017022449.1 3692 Intron XP_016877938.1

View Full Product Details