Product Details

SNP ID
rs115357955
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43189210 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTTTTTCATAGGAAACCCAGAA[A/G]TTCTGTGAACAAGTCCATGCTGCCA
Phenotype
MIM: 607089 MIM: 177070
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCNDBP1 PubMed Links

Gene Details

Gene
CCNDBP1
Gene Name
cyclin D1 binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012142.4 492 Silent Mutation AAA,AAG K87K NP_036274.3
XM_006720448.1 492 Silent Mutation AAA,AAG K131K XP_006720511.1
Gene
EPB42
Gene Name
erythrocyte membrane protein band 4.2
There are no transcripts associated with this gene.

Gene
TMEM62
Gene Name
transmembrane protein 62
There are no transcripts associated with this gene.

View Full Product Details