Product Details

SNP ID
rs117160809
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:74818085 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATAGGACGAGGGTGATAGGTGGGT[C/T]GGGGGAATGGAGGGCAGAAACTGTA
Phenotype
MIM: 609585 MIM: 609548
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CPLX3 PubMed Links

Gene Details

Gene
CPLX3
Gene Name
complexin 3
There are no transcripts associated with this gene.

Gene
LMAN1L
Gene Name
lectin, mannose binding 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021819.2 Intron NP_068591.2

View Full Product Details