Product Details

SNP ID
rs117581267
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:24986494 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGTCTCTGTTCTTCTGGTAGAAA[A/T]GTTAGTGTATTTATAACCTCTCAGA
Phenotype
MIM: 600162 MIM: 182279
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PWAR5 PubMed Links

Gene Details

Gene
PWAR5
Gene Name
Prader Willi/Angelman region RNA 5
There are no transcripts associated with this gene.

Gene
PWARSN
Gene Name
Prader Willi/Angelman region RNA, SNRPN neighbor
There are no transcripts associated with this gene.

Gene
SNORD107
Gene Name
small nucleolar RNA, C/D box 107
There are no transcripts associated with this gene.

Gene
SNORD108
Gene Name
small nucleolar RNA, C/D box 108
There are no transcripts associated with this gene.

Gene
SNORD64
Gene Name
small nucleolar RNA, C/D box 64
There are no transcripts associated with this gene.

Gene
SNRPN
Gene Name
small nuclear ribonucleoprotein polypeptide N
There are no transcripts associated with this gene.

Gene
SNURF
Gene Name
SNRPN upstream reading frame
There are no transcripts associated with this gene.

View Full Product Details