Product Details

SNP ID
rs137947373
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:65446886 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGAGTTCTACACAGGTCAAAATT[A/G]TATCACTTTTAGAGCAGCAATACGT
Phenotype
MIM: 606819
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DPP8 PubMed Links

Gene Details

Gene
DPP8
Gene Name
dipeptidyl peptidase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011521732.2 4010 UTR 3 XP_011520034.1
XM_017022378.1 4010 UTR 3 XP_016877867.1
XM_017022380.1 4010 UTR 3 XP_016877869.1

View Full Product Details