Product Details

SNP ID
rs138809807
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43777623 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGCAGGAAAGTTGCAGGGCAGA[A/G]AGCCAGAACCCAGTCCTAGGTCTCA
Phenotype
MIM: 609885 MIM: 602046 MIM: 605054
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ELL3 PubMed Links

Gene Details

Gene
ELL3
Gene Name
elongation factor for RNA polymerase II 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025165.2 Intron NP_079441.1
Gene
PDIA3
Gene Name
protein disulfide isomerase family A member 3
There are no transcripts associated with this gene.

Gene
SERF2
Gene Name
small EDRK-rich factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018108.3 Intron NP_001018118.1
NM_001199875.1 Intron NP_001186804.1
NM_001199876.1 Intron NP_001186805.1
NM_001199877.1 Intron NP_001186806.1
NM_001199878.1 Intron NP_001186807.1

View Full Product Details