Product Details

SNP ID
rs138814687
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40569365 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGCTGGACCGCCGCGGATGGCTC[A/C]GGGTTCTTGGACATTGGCGCTACGA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C15orf57 PubMed Links

Gene Details

Gene
C15orf57
Gene Name
chromosome 15 open reading frame 57
There are no transcripts associated with this gene.

Gene
RPUSD2
Gene Name
RNA pseudouridylate synthase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286407.1 73 Silent Mutation AGG,CGG R10R NP_001273336.1
NM_152260.2 73 Silent Mutation AGG,CGG R10R NP_689473.1

View Full Product Details