Product Details

SNP ID
rs138828358
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43328509 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGTACCAAAGGAAAAGCAGTTCC[C/G]ACCACCTCCAAGGAGCAGGAGCTGT
Phenotype
MIM: 611246
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADAL PubMed Links

Gene Details

Gene
ADAL
Gene Name
adenosine deaminase like
There are no transcripts associated with this gene.

Gene
LCMT2
Gene Name
leucine carboxyl methyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014793.4 2114 Missense Mutation CGG,GGG R661G NP_055608.2

View Full Product Details