Product Details

SNP ID
rs138899257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:84604268 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCAGGCTTGGCTGCAAGAGCATC[A/G]GCCTGAAAGCAGTGAGGAGGCAGCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZSCAN2 PubMed Links

Gene Details

Gene
ZSCAN2
Gene Name
zinc finger and SCAN domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007072.1 567 Missense Mutation CAG,CGG Q114R NP_001007073.1
NM_017894.5 567 Missense Mutation CAG,CGG Q114R NP_060364.4
NM_181877.3 567 Missense Mutation CAG,CGG Q114R NP_870992.2
XM_017022392.1 567 Missense Mutation CAG,CGG Q114R XP_016877881.1
XM_017022393.1 567 Missense Mutation CAG,CGG Q114R XP_016877882.1

View Full Product Details