Product Details

SNP ID
rs140227231
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:99568741 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTTTTAGTCTGGCAAGAAGAAT[A/G]TAGTGTAATATACGATGATGATGGG
Phenotype
MIM: 600660
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MEF2A PubMed Links

Gene Details

Gene
MEF2A
Gene Name
myocyte enhancer factor 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130926.2 1887 Intron NP_001124398.1
NM_001130927.2 1887 Intron NP_001124399.1
NM_001130928.2 1887 Intron NP_001124400.1
NM_001171894.2 1887 Intron NP_001165365.1
NM_001319206.1 1887 Intron NP_001306135.1
NM_005587.3 1887 Intron NP_005578.2
XM_005254914.2 1887 Intron XP_005254971.1
XM_005254915.2 1887 Intron XP_005254972.1
XM_005254916.3 1887 Intron XP_005254973.1
XM_011521571.2 1887 Intron XP_011519873.1
XM_011521572.2 1887 Intron XP_011519874.1
XM_011521573.2 1887 Intron XP_011519875.1
XM_011521576.2 1887 Intron XP_011519878.1
XM_011521577.2 1887 Intron XP_011519879.1
XM_011521578.2 1887 Intron XP_011519880.1
XM_011521579.2 1887 Intron XP_011519881.1
XM_011521581.2 1887 Intron XP_011519883.1
XM_011521582.2 1887 Intron XP_011519884.1
XM_011521583.2 1887 Intron XP_011519885.1
XM_011521585.2 1887 Intron XP_011519887.1
XM_011521586.2 1887 Intron XP_011519888.1
XM_011521587.2 1887 Intron XP_011519889.1
XM_011521590.2 1887 Intron XP_011519892.1
XM_017022190.1 1887 Intron XP_016877679.1
XM_017022191.1 1887 Intron XP_016877680.1
XM_017022192.1 1887 Intron XP_016877681.1
XM_017022193.1 1887 Intron XP_016877682.1
XM_017022194.1 1887 Intron XP_016877683.1
XM_017022195.1 1887 Intron XP_016877684.1
XM_017022196.1 1887 Intron XP_016877685.1
XM_017022197.1 1887 UTR 5 XP_016877686.1
XM_017022198.1 1887 Intron XP_016877687.1
XM_017022199.1 1887 Intron XP_016877688.1
XM_017022200.1 1887 Intron XP_016877689.1
XM_017022201.1 1887 Intron XP_016877690.1
XM_017022202.1 1887 Intron XP_016877691.1
XM_017022203.1 1887 Intron XP_016877692.1
XM_017022204.1 1887 Intron XP_016877693.1

View Full Product Details