Product Details

SNP ID
rs140723359
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:82043332 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACACGGAGCAGTCGCGGCTGCCT[C/T]TACGCCGAAGCCCGGAGGAAGAGGA
Phenotype
MIM: 611008
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MEX3B PubMed Links

Gene Details

Gene
MEX3B
Gene Name
mex-3 RNA binding family member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032246.4 1976 Missense Mutation AAA,AGA K513R NP_115622.2

View Full Product Details