Product Details

SNP ID
rs142170144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:73053893 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTTTCCCTGATTTATATGAAAT[A/G]ATATTTAAAATCACACTGCAAGGTA
Phenotype
MIM: 601907
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NEO1 PubMed Links

Gene Details

Gene
NEO1
Gene Name
neogenin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172623.1 Intron NP_001166094.1
NM_001172624.1 Intron NP_001166095.1
NM_002499.3 Intron NP_002490.2
XM_005254408.1 Intron XP_005254465.1
XM_011521628.2 Intron XP_011519930.1
XM_011521629.2 Intron XP_011519931.1
XM_011521630.2 Intron XP_011519932.1
XM_011521632.2 Intron XP_011519934.1
XM_011521633.2 Intron XP_011519935.1
XM_017022231.1 Intron XP_016877720.1
XM_017022232.1 Intron XP_016877721.1
XM_017022233.1 Intron XP_016877722.1
XM_017022234.1 Intron XP_016877723.1
XM_017022235.1 Intron XP_016877724.1
XM_017022236.1 Intron XP_016877725.1
XM_017022237.1 Intron XP_016877726.1
XM_017022238.1 Intron XP_016877727.1

View Full Product Details