Product Details

SNP ID
rs142515948
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41504539 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCAGCCCCTAGGAGGGTCCATC[C/T]GGCCTCCTCCAACGACGAAGTCCAG
Phenotype
MIM: 147521 MIM: 151520
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ITPKA PubMed Links

Gene Details

Gene
ITPKA
Gene Name
inositol-trisphosphate 3-kinase A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002220.2 2319 Intron NP_002211.1
XM_011521522.2 2319 Intron XP_011519824.1
Gene
LTK
Gene Name
leukocyte receptor tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135685.1 2319 Missense Mutation CAG,CGG Q611R NP_001129157.1
NM_002344.5 2319 Missense Mutation CAG,CGG Q741R NP_002335.2
NM_206961.3 2319 Missense Mutation CAG,CGG Q680R NP_996844.1
XM_011521556.2 2319 Missense Mutation CAG,CGG Q757R XP_011519858.1
XM_011521557.2 2319 Missense Mutation CAG,CGG Q701R XP_011519859.1
XM_017022181.1 2319 Missense Mutation CAG,CGG Q762R XP_016877670.1
XM_017022182.1 2319 Missense Mutation CAG,CGG Q714R XP_016877671.1
XM_017022183.1 2319 Missense Mutation CAG,CGG Q677R XP_016877672.1

View Full Product Details