Product Details

SNP ID
rs142660193
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75725145 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGTACCTCCGGCCATCCTGCAC[A/G]GGCTACATAGATCATGACATCTCCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNM1P35 PubMed Links

Gene Details

Gene
DNM1P35
Gene Name
dynamin 1 pseudogene 35
There are no transcripts associated with this gene.

Gene
ODF3L1
Gene Name
outer dense fiber of sperm tails 3 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175881.3 378 Silent Mutation ACA,ACG T52T NP_787077.1
XM_006720414.3 378 Silent Mutation ACA,ACG T52T XP_006720477.1

View Full Product Details