Product Details

SNP ID
rs143154243
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:45094684 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCACTTTCTGGAAGTGCCGCTCG[C/T]AGATGTACTGGGGGCACAGGGGCAG
Phenotype
MIM: 606759
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DUOX2 PubMed Links

Gene Details

Gene
DUOX2
Gene Name
dual oxidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014080.4 4606 Missense Mutation TAC,TGC Y1468C NP_054799.4
XM_005254421.2 4606 Missense Mutation TAC,TGC Y1468C XP_005254478.1

View Full Product Details