Product Details
- SNP ID
-
rs144091405
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:59107633 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACAAACAACTGAAACCTACTGCTT[C/T]TGTCAAACCAGTACAGATGGAAAAG
- Phenotype
-
MIM: 602755
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCNB2
PubMed Links
Gene Details
- Gene
- CCNB2
- Gene Name
- cyclin B2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004701.3 |
|
Intron |
|
|
NP_004692.1 |
View Full Product Details