Product Details
- SNP ID
-
rs145009665
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:99255980 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCATGGCGTCCGAACTTTGTAAGA[C/T]GATCTCTGTGGCAAGGCTAGAAAAG
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HSP90B2P
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2292313] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HSP90B2P
- Gene Name
- heat shock protein 90 beta family member 2, pseudogene
There are no transcripts associated with this gene.
- Gene
- LRRC28
- Gene Name
- leucine rich repeat containing 28
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001284400.2 |
263 |
Missense Mutation |
ACG,ATG |
T8M |
NP_001271329.1 |
NM_001321675.1 |
263 |
Missense Mutation |
ACG,ATG |
T8M |
NP_001308604.1 |
NM_001321676.1 |
263 |
Missense Mutation |
ACG,ATG |
T8M |
NP_001308605.1 |
NM_001321677.1 |
263 |
Missense Mutation |
ACG,ATG |
T8M |
NP_001308606.1 |
NM_001321678.1 |
263 |
Intron |
|
|
NP_001308607.1 |
NM_001321679.1 |
263 |
UTR 5 |
|
|
NP_001308608.1 |
NM_001321680.1 |
263 |
UTR 5 |
|
|
NP_001308609.1 |
NM_144598.4 |
263 |
Missense Mutation |
ACG,ATG |
T8M |
NP_653199.2 |
XM_006720389.3 |
263 |
Missense Mutation |
ACG,ATG |
T8M |
XP_006720452.1 |
XM_011521218.2 |
263 |
Missense Mutation |
ACG,ATG |
T8M |
XP_011519520.1 |
XM_011521220.1 |
263 |
Missense Mutation |
ACG,ATG |
T8M |
XP_011519522.1 |
XM_011521221.2 |
263 |
Missense Mutation |
ACG,ATG |
T8M |
XP_011519523.1 |
XM_017021914.1 |
263 |
Missense Mutation |
ACG,ATG |
T8M |
XP_016877403.1 |
- Gene
- TTC23
- Gene Name
- tetratricopeptide repeat domain 23
There are no transcripts associated with this gene.
View Full Product Details