Product Details

SNP ID
rs145665583
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:98969618 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGACTGTTCCAGAATGATCGCCT[G/T]GGCGGCGGTGTCCATGCCCACATGC
Phenotype
MIM: 147370
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IGF1R PubMed Links

Gene Details

Gene
IGF1R
Gene Name
insulin like growth factor 1 receptor
There are no transcripts associated with this gene.

Gene
PGPEP1L
Gene Name
pyroglutamyl-peptidase I-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102612.2 479 Missense Mutation AAG,CAG K60Q NP_001096082.2
NM_001167902.1 479 Intron NP_001161374.1
XM_011521254.1 479 Missense Mutation AAG,CAG K151Q XP_011519556.1
XM_017021933.1 479 Missense Mutation AAG,CAG K6Q XP_016877422.1

View Full Product Details