Product Details

SNP ID
rs146617036
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41644208 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGCCTAACATTTCATTCAAAAT[A/G]TTACTATACAGGCCAGGTACAGTGA
Phenotype
MIM: 616061
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MGA PubMed Links

Gene Details

Gene
MGA
Gene Name
MGA, MAX dimerization protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080541.2 Intron NP_001074010.2
NM_001164273.1 Intron NP_001157745.1
XM_005254243.2 Intron XP_005254300.1
XM_005254244.3 Intron XP_005254301.1
XM_005254245.4 Intron XP_005254302.1
XM_005254246.2 Intron XP_005254303.1
XM_005254247.2 Intron XP_005254304.1
XM_005254248.2 Intron XP_005254305.1
XM_005254249.2 Intron XP_005254306.1
XM_005254252.2 Intron XP_005254309.1
XM_005254253.2 Intron XP_005254310.1
XM_005254254.3 Intron XP_005254311.1
XM_006720443.3 Intron XP_006720506.1
XM_006720445.3 Intron XP_006720508.1
XM_011521397.2 Intron XP_011519699.1
XM_011521398.2 Intron XP_011519700.1
XM_011521399.2 Intron XP_011519701.1
XM_011521400.2 Intron XP_011519702.1
XM_011521401.2 Intron XP_011519703.1
XM_017022028.1 Intron XP_016877517.1
XM_017022029.1 Intron XP_016877518.1
XM_017022030.1 Intron XP_016877519.1
XM_017022031.1 Intron XP_016877520.1
XM_017022032.1 Intron XP_016877521.1
XM_017022033.1 Intron XP_016877522.1

View Full Product Details