Product Details

SNP ID
rs147092381
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:76350579 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGATGCTGTCATAACACCCAAAG[A/G]GCAACTTGTACTTCTCAATTACAGT
Phenotype
MIM: 609481 MIM: 611611
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ISL2 PubMed Links

Gene Details

Gene
ISL2
Gene Name
ISL LIM homeobox 2
There are no transcripts associated with this gene.

Gene
SCAPER
Gene Name
S-phase cyclin A associated protein in the ER
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145923.1 Intron NP_001139395.1
NM_020843.2 Intron NP_065894.2
XM_005254417.2 Intron XP_005254474.1
XM_005254419.2 Intron XP_005254476.1
XM_011521650.1 Intron XP_011519952.1
XM_011521652.2 Intron XP_011519954.1
XM_011521653.2 Intron XP_011519955.1
XM_011521654.2 Intron XP_011519956.1
XM_011521656.2 Intron XP_011519958.1
XM_017022266.1 Intron XP_016877755.1
XM_017022267.1 Intron XP_016877756.1
XM_017022268.1 Intron XP_016877757.1
XM_017022269.1 Intron XP_016877758.1
XM_017022270.1 Intron XP_016877759.1
XM_017022271.1 Intron XP_016877760.1
XM_017022272.1 Intron XP_016877761.1
XM_017022273.1 Intron XP_016877762.1
XM_017022274.1 Intron XP_016877763.1
XM_017022275.1 Intron XP_016877764.1
XM_017022276.1 Intron XP_016877765.1
XM_017022277.1 Intron XP_016877766.1
XM_017022278.1 Intron XP_016877767.1
XM_017022279.1 Intron XP_016877768.1
XM_017022280.1 Intron XP_016877769.1
XM_017022281.1 Intron XP_016877770.1
XM_017022282.1 Intron XP_016877771.1
XM_017022283.1 Intron XP_016877772.1
XM_017022284.1 Intron XP_016877773.1

View Full Product Details