Product Details

SNP ID
rs147791097
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:45094269 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCAGCTGAACACCCCGATCTTGC[A/G]CACCTGTCAGGAGATTGGAGAGAGA
Phenotype
MIM: 606759
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUOX2 PubMed Links

Gene Details

Gene
DUOX2
Gene Name
dual oxidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014080.4 4731 Missense Mutation CGC,TGC R1510C NP_054799.4
XM_005254421.2 4731 Missense Mutation CGC,TGC R1510C XP_005254478.1

View Full Product Details