Product Details

SNP ID
rs149257811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41394926 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAACTATACATCTGATTCGTCCTC[C/T]GGAAGAAATCTGTGTCCTCCTTGAT
Phenotype
MIM: 606934
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NDUFAF1 PubMed Links

Gene Details

Gene
NDUFAF1
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016013.3 1020 Missense Mutation CAG,CGG Q231R NP_057097.2
XM_006720555.2 1020 Missense Mutation CAG,CGG Q231R XP_006720618.1
XM_011521658.1 1020 Missense Mutation CAG,CGG Q231R XP_011519960.1
XM_011521659.2 1020 Missense Mutation CAG,CGG Q231R XP_011519961.1
XM_017022293.1 1020 Intron XP_016877782.1

View Full Product Details