Product Details

SNP ID
rs149258367
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78173714 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCTCTTCGATGGCCTGGTACAC[A/G]GCCTCATCCTTCTTCTCTATGATCT
Phenotype
MIM: 614362 MIM: 601149
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACSBG1 PubMed Links

Gene Details

Gene
ACSBG1
Gene Name
acyl-CoA synthetase bubblegum family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199377.1 2162 Silent Mutation GCC,GCT A652A NP_001186306.1
NM_015162.4 2162 Silent Mutation GCC,GCT A656A NP_055977.3
XM_011521390.2 2162 Silent Mutation GCC,GCT A634A XP_011519692.1
XM_011521391.2 2162 Silent Mutation GCC,GCT A585A XP_011519693.2
XM_017022025.1 2162 Silent Mutation GCC,GCT A656A XP_016877514.1
XM_017022026.1 2162 Silent Mutation GCC,GCT A664A XP_016877515.1
Gene
IDH3A
Gene Name
isocitrate dehydrogenase 3 (NAD(+)) alpha
There are no transcripts associated with this gene.

View Full Product Details