Product Details

SNP ID
rs149264316
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:96785090 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGAAGATAAACAGGAGAAGTGT[C/T]CTGTTTCACCCATATTGCTGGTCCT
Phenotype
MIM: 613948
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPATA8 PubMed Links

Gene Details

Gene
SPATA8
Gene Name
spermatogenesis associated 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304804.1 385 Silent Mutation GTC,GTT V58V NP_001291733.1
NM_001304805.1 385 Silent Mutation GTC,GTT V58V NP_001291734.1
NM_001304806.1 385 Silent Mutation GTC,GTT V58V NP_001291735.1
NM_173499.4 385 Silent Mutation GTC,GTT V97V NP_775770.1
XM_011521265.2 385 Silent Mutation GTC,GTT V58V XP_011519567.1
Gene
SPATA8-AS1
Gene Name
SPATA8 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details