Product Details

SNP ID
rs149327023
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:42529758 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCTGGGAAATCTAAAAGACATGG[C/T]CCTGAACATAGGCAATGAGATTGAT
Phenotype
MIM: 602534
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC57 PubMed Links

Gene Details

Gene
LRRC57
Gene Name
leucine rich repeat containing 57
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153260.2 1010 Intron NP_694992.2
XM_011521423.2 1010 Intron XP_011519725.1
XM_011521424.2 1010 Intron XP_011519726.1
Gene
SNAP23
Gene Name
synaptosome associated protein 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003825.3 1010 Missense Mutation GCC,GTC A170V NP_003816.2
NM_130798.2 1010 Missense Mutation GCC,GTC A117V NP_570710.1
XM_006720725.2 1010 Missense Mutation GCC,GTC A181V XP_006720788.1
XM_017022692.1 1010 Missense Mutation GCC,GTC A181V XP_016878181.1
XM_017022693.1 1010 Intron XP_016878182.1
XM_017022694.1 1010 Intron XP_016878183.1

View Full Product Details