Product Details

SNP ID
rs149344478
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:23566184 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCACTGAGGGTGGCGTTTCGCC[A/G]CCTGGGGCCTCTGCAGGTGGAGGCC
Phenotype
MIM: 603856
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR4508 PubMed Links

Gene Details

Gene
MIR4508
Gene Name
microRNA 4508
There are no transcripts associated with this gene.

Gene
MKRN3
Gene Name
makorin ring finger protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005664.3 878 Silent Mutation CCA,CCG P134P NP_005655.1

View Full Product Details