Product Details

SNP ID
rs149611843
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:51058044 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGTGGGTCCTGGGCGTCAGGTGC[C/T]GCTGCACCAGTTCATGCACCAGGTC
Phenotype
MIM: 616438
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PIRC66 PubMed Links

Gene Details

Gene
PIRC66
Gene Name
piwi-interacting RNA cluster 66
There are no transcripts associated with this gene.

Gene
TNFAIP8L3
Gene Name
TNF alpha induced protein 8 like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001311175.1 915 Missense Mutation CAG,CGG Q151R NP_001298104.1
NM_207381.3 915 Missense Mutation CAG,CGG Q239R NP_997264.2
XM_011521545.2 915 Missense Mutation CAG,CGG Q165R XP_011519847.1
XM_017022168.1 915 Missense Mutation CAG,CGG Q150R XP_016877657.1
XM_017022169.1 915 Missense Mutation CAG,CGG Q105R XP_016877658.1

View Full Product Details