Product Details

SNP ID
rs150595531
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41503799 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGATCAAAAGCTGGAAGTGGCTGG[A/G]CCCTTCCCTGGGAGGCCTGGGCTGG
Phenotype
MIM: 147521 MIM: 151520
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITPKA PubMed Links

Gene Details

Gene
ITPKA
Gene Name
inositol-trisphosphate 3-kinase A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002220.2 3033 Intron NP_002211.1
XM_011521522.2 3033 Intron XP_011519824.1
Gene
LTK
Gene Name
leukocyte receptor tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135685.1 3033 UTR 3 NP_001129157.1
NM_002344.5 3033 UTR 3 NP_002335.2
NM_206961.3 3033 UTR 3 NP_996844.1
XM_011521556.2 3033 Intron XP_011519858.1
XM_011521557.2 3033 Intron XP_011519859.1
XM_017022181.1 3033 UTR 3 XP_016877670.1
XM_017022182.1 3033 Intron XP_016877671.1
XM_017022183.1 3033 Intron XP_016877672.1

View Full Product Details