Product Details

SNP ID
rs137902355
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2344831 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATATTTGTCTAAACCAAATCACTC[A/G]CTTTAAAATAGTTATTTTGCTTAGG
Phenotype
MIM: 601615
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ABCA17P PubMed Links

Gene Details

Gene
ABCA17P
Gene Name
ATP binding cassette subfamily A member 17, pseudogene
There are no transcripts associated with this gene.

Gene
ABCA3
Gene Name
ATP binding cassette subfamily A member 3
There are no transcripts associated with this gene.

View Full Product Details