Product Details

SNP ID
rs139772883
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:57358882 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTGCTGTGGCGCTTCAAGCAAC[C/T]GAGGCAGGTGAGGCTGGGGAGCAGG
Phenotype
MIM: 602957
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCL22 PubMed Links

Gene Details

Gene
CCL22
Gene Name
C-C motif chemokine ligand 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002990.4 120 Silent Mutation ACC,ACT T22T NP_002981.2
XM_017023531.1 120 Silent Mutation ACC,ACT T22T XP_016879020.1

View Full Product Details