Product Details
- SNP ID
-
rs140142659
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:31394330 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTTTGGGCAGAGCGTGGTGCAGTT[C/T]GGTGGATCTCGGTAGGCCCCACTCA
- Phenotype
-
MIM: 602453
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ITGAD
PubMed Links
Gene Details
- Gene
- ITGAD
- Gene Name
- integrin subunit alpha D
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001318185.1 |
253 |
Silent Mutation |
TTC,TTT |
F42F |
NP_001305114.1 |
NM_005353.2 |
253 |
Silent Mutation |
TTC,TTT |
F42F |
NP_005344.2 |
XM_011545835.2 |
253 |
Missense Mutation |
TTC,TTT |
F42F |
XP_011544137.1 |
XM_011545836.2 |
253 |
Missense Mutation |
TTC,TTT |
F42F |
XP_011544138.1 |
XM_011545837.2 |
253 |
Missense Mutation |
TTC,TTT |
F42F |
XP_011544139.1 |
XM_011545838.2 |
253 |
Missense Mutation |
TTC,TTT |
F42F |
XP_011544140.1 |
XM_011545839.2 |
253 |
Silent Mutation |
TTC,TTT |
F42F |
XP_011544141.1 |
XM_011545841.2 |
253 |
Missense Mutation |
TTC,TTT |
F42F |
XP_011544143.1 |
XM_011545842.2 |
253 |
Missense Mutation |
TTC,TTT |
F42F |
XP_011544144.1 |
XM_011545843.2 |
253 |
Missense Mutation |
TTC,TTT |
F42F |
XP_011544145.1 |
XM_011545844.2 |
253 |
Missense Mutation |
TTC,TTT |
F42F |
XP_011544146.1 |
XM_011545845.2 |
253 |
Missense Mutation |
TTC,TTT |
F42F |
XP_011544147.1 |
XM_011545846.2 |
253 |
Missense Mutation |
TTC,TTT |
F42F |
XP_011544148.1 |
XM_011545847.2 |
253 |
Missense Mutation |
TTC,TTT |
F42F |
XP_011544149.1 |
XM_011545848.2 |
253 |
Missense Mutation |
TTC,TTT |
F42F |
XP_011544150.1 |
XM_017023215.1 |
253 |
Missense Mutation |
TTC,TTT |
F42F |
XP_016878704.1 |
View Full Product Details