Product Details

SNP ID
rs140415359
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31216933 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGGATGCGCTCAAAGCCCTCGCG[C/G]AGCGGCACCGTCCCCAGCTTCATCT
Phenotype
MIM: 615700 MIM: 613288
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PYDC1 PubMed Links

Gene Details

Gene
PYDC1
Gene Name
pyrin domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152901.3 161 Silent Mutation CTC,CTG L32L NP_690865.1
Gene
TRIM72
Gene Name
tripartite motif containing 72
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008274.3 161 Intron NP_001008275.2
XM_017023254.1 161 Intron XP_016878743.1

View Full Product Details