Product Details

SNP ID
rs140418377
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67609563 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTGATTACTGTCTCTTTAAGAC[C/T]CTTTTATCTGTATCAGAGTTTCTCA
Phenotype
MIM: 604167
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTCF PubMed Links

Gene Details

Gene
CTCF
Gene Name
CCCTC-binding factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001191022.1 Intron NP_001177951.1
NM_006565.3 Intron NP_006556.1
XM_005255775.3 Intron XP_005255832.1
XM_017022868.1 Intron XP_016878357.1
XM_017022869.1 Intron XP_016878358.1

View Full Product Details