Product Details

SNP ID
rs140563705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:21953444 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATCATGAAGCTACTAACCAGAGC[C/T]GGCTCTTTCTCGGTGAGCTCAGGTG
Phenotype
MIM: 191329
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDZD9 PubMed Links

Gene Details

Gene
PDZD9
Gene Name
PDZ domain containing 9
There are no transcripts associated with this gene.

Gene
UQCRC2
Gene Name
ubiquinol-cytochrome c reductase core protein II
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003366.3 381 Silent Mutation GCC,GCT A7A NP_003357.2

View Full Product Details