Product Details

SNP ID
rs140814462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:70253289 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACCTGGGGAACTTGACACAGGCA[C/T]GTGATCTTGCCAGCCTCATTGTCCA
Phenotype
MIM: 601065 MIM: 606490
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AARS PubMed Links

Gene Details

Gene
AARS
Gene Name
alanyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001605.2 2810 Silent Mutation ACA,ACG T900T NP_001596.2
Gene
EXOSC6
Gene Name
exosome component 6
There are no transcripts associated with this gene.

Gene
LOC107984138
Gene Name
serine/threonine-protein kinase SMG1-like
There are no transcripts associated with this gene.

View Full Product Details