Product Details
- SNP ID
-
rs140972426
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:49281217 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAACACTCACCTGGTCGAAGTAGAT[G/T]ATCATGGTGCGATTACTCATCTCGG
- Phenotype
-
MIM: 600432
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CBLN1
PubMed Links
Gene Details
- Gene
- CBLN1
- Gene Name
- cerebellin 1 precursor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004352.3 |
615 |
Silent Mutation |
ATA,ATC |
I83I |
NP_004343.1 |
View Full Product Details