Product Details

SNP ID
rs141134476
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:574305 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGGCAGGAGCCCGAGGAGAGCCT[A/G]GGCCGCTTCCTGGAGAGCCTGGGTG
Phenotype
MIM: 605754
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC107987233 PubMed Links

Gene Details

Gene
LOC107987233
Gene Name
uncharacterized LOC107987233
There are no transcripts associated with this gene.

Gene
NHLRC4
Gene Name
NHL repeat containing 4
There are no transcripts associated with this gene.

Gene
PIGQ
Gene Name
phosphatidylinositol glycan anchor biosynthesis class Q
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004204.3 369 Silent Mutation CTA,CTG L77L NP_004195.2
NM_148920.2 369 Silent Mutation CTA,CTG L77L NP_683721.1
Gene
PRR35
Gene Name
proline rich 35
There are no transcripts associated with this gene.

View Full Product Details