Product Details

SNP ID
rs143500674
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:82148841 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTTTCTGGCAAACTTTTTCCCAA[C/T]TGTCCCCACCAAGGTCTCATATCTG
Phenotype
MIM: 605500
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MPHOSPH6 PubMed Links

Gene Details

Gene
MPHOSPH6
Gene Name
M-phase phosphoprotein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005792.2 536 Missense Mutation ATT,GTT I125V NP_005783.2
XM_011522808.2 536 Missense Mutation ATT,GTT I107V XP_011521110.1
XM_017022852.1 536 Missense Mutation ATT,GTT I107V XP_016878341.1

View Full Product Details