Product Details
- SNP ID
-
rs144460906
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:66995700 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGCAAGGAAAACAGATGCATGTGG[A/G]GAAGACACCCATCCAAGTCTTTTTA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CES4A
PubMed Links
Gene Details
- Gene
- CES4A
- Gene Name
- carboxylesterase 4A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001190201.1 |
312 |
Intron |
|
|
NP_001177130.1 |
NM_001190202.1 |
312 |
Intron |
|
|
NP_001177131.1 |
NM_001318506.1 |
312 |
Intron |
|
|
NP_001305435.1 |
NM_173815.6 |
312 |
Missense Mutation |
GAG,GGG |
E44G |
NP_776176.5 |
XM_005255896.4 |
312 |
Missense Mutation |
GAG,GGG |
E67G |
XP_005255953.1 |
XM_011523021.2 |
312 |
Missense Mutation |
GAG,GGG |
E67G |
XP_011521323.1 |
XM_011523023.2 |
312 |
Missense Mutation |
GAG,GGG |
E67G |
XP_011521325.1 |
XM_011523024.2 |
312 |
Missense Mutation |
GAG,GGG |
E67G |
XP_011521326.1 |
XM_011523025.2 |
312 |
Missense Mutation |
GAG,GGG |
E67G |
XP_011521327.1 |
XM_011523026.2 |
312 |
Missense Mutation |
GAG,GGG |
E67G |
XP_011521328.1 |
XM_011523027.2 |
312 |
Missense Mutation |
GAG,GGG |
E67G |
XP_011521329.1 |
XM_011523028.2 |
312 |
Missense Mutation |
GAG,GGG |
E55G |
XP_011521330.1 |
XM_011523029.2 |
312 |
Missense Mutation |
GAG,GGG |
E67G |
XP_011521331.1 |
XM_011523030.2 |
312 |
Missense Mutation |
GAG,GGG |
E67G |
XP_011521332.1 |
XM_017023160.1 |
312 |
Missense Mutation |
GAG,GGG |
E67G |
XP_016878649.1 |
XM_017023161.1 |
312 |
Missense Mutation |
GAG,GGG |
E67G |
XP_016878650.1 |
XM_017023162.1 |
312 |
Missense Mutation |
GAG,GGG |
E67G |
XP_016878651.1 |
XM_017023163.1 |
312 |
Missense Mutation |
GAG,GGG |
E67G |
XP_016878652.1 |
XM_017023164.1 |
312 |
Missense Mutation |
GAG,GGG |
E67G |
XP_016878653.1 |
XM_017023165.1 |
312 |
Missense Mutation |
GAG,GGG |
E67G |
XP_016878654.1 |
XM_017023166.1 |
312 |
Missense Mutation |
GAG,GGG |
E67G |
XP_016878655.1 |
XM_017023167.1 |
312 |
Intron |
|
|
XP_016878656.1 |
XM_017023168.1 |
312 |
Intron |
|
|
XP_016878657.1 |
View Full Product Details